Product Details

SNP ID
rs3917637
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:94529198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGTCACCAAAATGTATTATAAG[A/C]GAAAAAGATACGTTGTTGTAAGCCA
Phenotype
MIM: 134390
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
F3 PubMed Links

Gene Details

Gene
F3
Gene Name
coagulation factor III, tissue factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178096.1 2371 UTR 3 NP_001171567.1
NM_001993.4 2371 UTR 3 NP_001984.1

View Full Product Details