Product Details

SNP ID
rs3917837
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:169591917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCCATAGAAAACTTCAAAGCCT[A/G]AAATCAAATTGGGAGAAGCTGAGGA
Phenotype
MIM: 612309 MIM: 173610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
F5 PubMed Links

Gene Details

Gene
F5
Gene Name
coagulation factor V
There are no transcripts associated with this gene.

Gene
SELP
Gene Name
selectin P
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003005.3 Intron NP_002996.2
XM_005245435.1 Intron XP_005245492.1
XM_005245436.3 Intron XP_005245493.1
XM_005245438.1 Intron XP_005245495.1
XM_005245439.1 Intron XP_005245496.1
XM_005245440.1 Intron XP_005245497.1

View Full Product Details