Product Details

SNP ID
rs17421358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:140862135 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCCTGTCACTTCACTAAGGATAA[C/T]AGACCACAGATATTCTGCCAAGCAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF150 PubMed Links

Gene Details

Gene
RNF150
Gene Name
ring finger protein 150
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020724.1 6981 Intron NP_065775.1
XM_005263150.4 6981 UTR 3 XP_005263207.1
XM_011532147.2 6981 UTR 3 XP_011530449.1
XM_011532148.2 6981 UTR 3 XP_011530450.1
XM_017008475.1 6981 UTR 3 XP_016863964.1
XM_017008476.1 6981 UTR 3 XP_016863965.1

View Full Product Details