Product Details

SNP ID
rs16832105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:199957584 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCATGGCCATTTGTATTTATAAGA[C/G]CAATCAGTTGTAAATCAAATGTTTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C2orf47 PubMed Links

Gene Details

Gene
C2orf47
Gene Name
chromosome 2 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024520.2 Intron NP_078796.2
XM_005246852.2 Intron XP_005246909.1
Gene
TYW5
Gene Name
tRNA-yW synthesizing protein 5
There are no transcripts associated with this gene.

View Full Product Details