Product Details

SNP ID
rs17786967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:149136718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCATAAGGATTGTCAGGCAGGC[A/C]CATTTAGCAAGTGCTGGATCATCAT
Phenotype
MIM: 606118
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HPS3 PubMed Links

Gene Details

Gene
HPS3
Gene Name
HPS3, biogenesis of lysosomal organelles complex 2 subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308258.1 Intron NP_001295187.1
NM_032383.4 Intron NP_115759.2
XM_005247834.3 Intron XP_005247891.1
XM_017007323.1 Intron XP_016862812.1

View Full Product Details