Product Details

SNP ID
rs17852745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219162315 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACTTACCAGGAATCGTGTTCTC[C/T]AGTAGGAAGCCTGAGAGTCCAGCCA
Phenotype
MIM: 611290
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNPPD1 PubMed Links

Gene Details

Gene
CNPPD1
Gene Name
cyclin Pas1/PHO80 domain containing 1
There are no transcripts associated with this gene.

Gene
NHEJ1
Gene Name
non-homologous end joining factor 1
There are no transcripts associated with this gene.

Gene
SLC23A3
Gene Name
solute carrier family 23 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144889.1 1281 Silent Mutation CTA,CTG L507L NP_001138361.1
NM_001144890.1 1281 Silent Mutation CTA,CTG L515L NP_001138362.1
NM_144712.4 1281 Silent Mutation CTA,CTG L390L NP_653313.3

View Full Product Details