Product Details

SNP ID
rs16833132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:122423817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGCCAATCTTTTTATCTGTGAC[A/G]ATAATGCCAAAAATGACTCAGAGCC
Phenotype
MIM: 600686
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KPNA1 PubMed Links

Gene Details

Gene
KPNA1
Gene Name
karyopherin subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002264.3 6570 UTR 3 NP_002255.3
XM_005247437.3 6570 UTR 3 XP_005247494.1
XM_005247439.3 6570 UTR 3 XP_005247496.1
XM_011512795.2 6570 UTR 3 XP_011511097.1
XM_011512797.2 6570 UTR 3 XP_011511099.1
XM_017006363.1 6570 UTR 3 XP_016861852.1
XM_017006364.1 6570 UTR 3 XP_016861853.1
XM_017006365.1 6570 UTR 3 XP_016861854.1
Gene
LOC102723582
Gene Name
uncharacterized LOC102723582
There are no transcripts associated with this gene.

Gene
WDR5B
Gene Name
WD repeat domain 5B
There are no transcripts associated with this gene.

View Full Product Details