Product Details

SNP ID
rs16875751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:24528573 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGTCCTACACAATTTAAGTGTCA[A/G]AAGTCCCAAATCCGTATAAGAAAGC
Phenotype
MIM: 603403
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHX15 PubMed Links

Gene Details

Gene
DHX15
Gene Name
DEAH-box helicase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001358.2 Intron NP_001349.2
Gene
MIR573
Gene Name
microRNA 573
There are no transcripts associated with this gene.

View Full Product Details