Product Details

SNP ID
rs16995553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35264612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAACTTGTTAGAAGTTGCTTCC[A/C]ATCATATTGACAGTACTTCTCTTGA
Phenotype
MIM: 604702
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HMGXB4 PubMed Links

Gene Details

Gene
HMGXB4
Gene Name
HMG-box containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003681.2 Intron NP_001003681.1
XM_006724100.3 Intron XP_006724163.1
XM_006724101.3 Intron XP_006724164.1
XM_006724102.1 Intron XP_006724165.1

View Full Product Details