Product Details

SNP ID
rs17844384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141100713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTTGCTTCTCTTTGTTTTTCTG[A/G]GAGGGTCTCTGGCTGGGTCCGAGTC
Phenotype
MIM: 606328 MIM: 606329
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHB2 PubMed Links

Gene Details

Gene
PCDHB2
Gene Name
protocadherin beta 2
There are no transcripts associated with this gene.

Gene
PCDHB3
Gene Name
protocadherin beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018937.4 472 Missense Mutation AGA,GGA R22G NP_061760.1

View Full Product Details