Product Details
- SNP ID
-
rs16967294
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:75736525 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCAGAGTCACAACACAGAAGCTC[A/G]CCTGCACACAGAGGCATTTGCACCA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DNM1P35
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs563457191] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DNM1P35
- Gene Name
- dynamin 1 pseudogene 35
There are no transcripts associated with this gene.
- Gene
- ODF3L1
- Gene Name
- outer dense fiber of sperm tails 3 like 1
There are no transcripts associated with this gene.
View Full Product Details