Product Details

SNP ID
rs17032290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:102253571 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGCTTTTTTAGAATACATGTTCA[C/T]TGAAGAAAATTGGAAAGCATTAAAA
Phenotype
MIM: 608732
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC39A8 PubMed Links

Gene Details

Gene
SLC39A8
Gene Name
solute carrier family 39 member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135146.1 Intron NP_001128618.1
NM_001135147.1 Intron NP_001128619.1
NM_001135148.1 Intron NP_001128620.1
NM_022154.5 Intron NP_071437.3
XM_005263177.1 Intron XP_005263234.1
XM_011532181.2 Intron XP_011530483.1
XM_011532182.1 Intron XP_011530484.1
XM_017008540.1 Intron XP_016864029.1
XM_017008541.1 Intron XP_016864030.1
XM_017008542.1 Intron XP_016864031.1

View Full Product Details