Product Details

SNP ID
rs16921226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94963632 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGATGCTCAACATGTGTATCATC[C/T]GATGCAGCAATTACACTCAAAAGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CWC15 PubMed Links

Gene Details

Gene
CWC15
Gene Name
CWC15 spliceosome-associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016403.3 Intron NP_057487.2
XM_011542868.2 Intron XP_011541170.1
XM_017017916.1 Intron XP_016873405.1

View Full Product Details