Product Details

SNP ID
rs17291811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1843757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAGTGGAGGGAGGTTAGTGGGAA[A/G]ATCATCAAATCCAGTTTTCTTTTAC
Phenotype
MIM: 179835
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPA1 PubMed Links

Gene Details

Gene
RPA1
Gene Name
replication protein A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002945.3 Intron NP_002936.1

View Full Product Details