Product Details

SNP ID
rs35278805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43361124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTTCCCGTGCATGGATTTCTCC[A/G]TGCTTATTAAGGGCAGAGCTTTTAC
Phenotype
MIM: 609610
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAL PubMed Links

Gene Details

Gene
ADAL
Gene Name
adenosine deaminase like
There are no transcripts associated with this gene.

Gene
TUBGCP4
Gene Name
tubulin gamma complex associated protein 4
There are no transcripts associated with this gene.

Gene
ZSCAN29
Gene Name
zinc finger and SCAN domain containing 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152455.3 3095 Silent Mutation CAC,CAT H836H NP_689668.3
XM_006720401.3 3095 Silent Mutation CAC,CAT H836H XP_006720464.1
XM_011521266.2 3095 Silent Mutation CAC,CAT H836H XP_011519568.1
XM_017021949.1 3095 Intron XP_016877438.1

View Full Product Details