Product Details

SNP ID
rs17727765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:29249944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAGATATTATCCCTGGCAGACCA[C/T]TTCTATCATAAACCAATCCTCCCTC
Phenotype
MIM: 123610 MIM: 609356
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYBA1 PubMed Links

Gene Details

Gene
CRYBA1
Gene Name
crystallin beta A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005208.4 Intron NP_005199.2
XM_017024198.1 Intron XP_016879687.1
Gene
NUFIP2
Gene Name
NUFIP2, FMR1 interacting protein 2
There are no transcripts associated with this gene.

View Full Product Details