Product Details

SNP ID
rs16988468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:72584590 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCTTTTGGGATAGACTAGTCCCA[A/C]ATATTCAGAGACTATCTGGAAAGAT
Phenotype
MIM: 311870
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PHKA1 PubMed Links

Gene Details

Gene
PHKA1
Gene Name
phosphorylase kinase regulatory subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122670.1 Intron NP_001116142.1
NM_001172436.1 Intron NP_001165907.1
NM_002637.3 Intron NP_002628.2
XM_006724661.2 Intron XP_006724724.1

View Full Product Details