Product Details

SNP ID
rs17129464
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94093797 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCTGTCCTTTTCCCCTACTGGAA[C/T]AAATTTCATGAAATAAGGATGTGTC
Phenotype
MIM: 611320
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFI27L1 PubMed Links

Gene Details

Gene
IFI27L1
Gene Name
interferon alpha inducible protein 27 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145249.2 Intron NP_660292.1
NM_206949.2 Intron NP_996832.1
XM_005267314.4 Intron XP_005267371.1
XM_011536405.2 Intron XP_011534707.1
XM_011536406.1 Intron XP_011534708.1
XM_011536407.1 Intron XP_011534709.1
XM_017020961.1 Intron XP_016876450.1
XM_017020962.1 Intron XP_016876451.1
XM_017020963.1 Intron XP_016876452.1

View Full Product Details