Product Details

SNP ID
rs17483706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89708763 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGCACATTTCTTCCTCAGAAAAA[C/G]TAACAAGGAGAATGGTACATAGTAA
Phenotype
MIM: 605498
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIF20B PubMed Links

Gene Details

Gene
KIF20B
Gene Name
kinesin family member 20B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284259.1 Intron NP_001271188.1
NM_016195.3 Intron NP_057279.2

View Full Product Details