Product Details

SNP ID
rs17717320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:147826351 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTATTTGAGAAAGCAACATTTTGT[C/G]TCTAGACCTAAATTTTCTTATCTGA
Phenotype
MIM: 167790
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPINK1 PubMed Links

Gene Details

Gene
SPINK1
Gene Name
serine peptidase inhibitor, Kazal type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003122.4 Intron NP_003113.2

View Full Product Details