Product Details

SNP ID
rs17183668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:4440075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAGCTCTGGTGAAGAACGCACT[C/T]GAGCCAAGTGAACGCCATGTTCATA
Phenotype
MIM: 134921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF6 PubMed Links

Gene Details

Gene
FGF6
Gene Name
fibroblast growth factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020996.2 2001 Intron NP_066276.2
XM_017018995.1 2001 UTR 3 XP_016874484.1
XM_017018996.1 2001 Intron XP_016874485.1

View Full Product Details