Product Details

SNP ID
rs7802977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:32586388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAGTTTTTCAAGGAACACTGGC[C/T]GTTGTATTGTTGGCAGGGGCCCCAA
Phenotype
MIM: 612927
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AVL9 PubMed Links

Gene Details

Gene
AVL9
Gene Name
AVL9 cell migration associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015060.2 4489 UTR 3 NP_055875.1
XM_005249668.2 4489 UTR 3 XP_005249725.1
XM_005249669.2 4489 UTR 3 XP_005249726.1
XM_005249670.2 4489 UTR 3 XP_005249727.1
XM_005249671.4 4489 Intron XP_005249728.1
XM_011515227.2 4489 UTR 3 XP_011513529.1
XM_017011891.1 4489 UTR 3 XP_016867380.1
XM_017011892.1 4489 UTR 3 XP_016867381.1
Gene
DPY19L1P1
Gene Name
DPY19L1 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details