Product Details

SNP ID
rs36058408
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:23747579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTTTTGACCTTTTCAGTGATT[C/T]AAAATTTTCCTTTAAGTGAAGGAAT
Phenotype
MIM: 600786
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TCEB3 PubMed Links

Gene Details

Gene
TCEB3
Gene Name
transcription elongation factor B subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003198.2 Intron NP_003189.2

View Full Product Details