Product Details

SNP ID
rs16940881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110500013 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTTAATTATTCAAGTATATGGAT[C/G]TTAATTATCTTACATTCAAAATTCT
Phenotype
MIM: 608368 MIM: 606932
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM216A PubMed Links

Gene Details

Gene
FAM216A
Gene Name
family with sequence similarity 216 member A
There are no transcripts associated with this gene.

Gene
RAD9B
Gene Name
RAD9 checkpoint clamp component B
There are no transcripts associated with this gene.

Gene
VPS29
Gene Name
VPS29, retromer complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282150.1 Intron NP_001269079.1
NM_001282151.1 Intron NP_001269080.1
NM_016226.4 Intron NP_057310.1
NM_057180.2 Intron NP_476528.1
XM_006719459.3 Intron XP_006719522.1
XM_006719460.3 Intron XP_006719523.1
XM_011538484.2 Intron XP_011536786.1
XM_017019457.1 Intron XP_016874946.1
XM_017019458.1 Intron XP_016874947.1
XM_017019459.1 Intron XP_016874948.1

View Full Product Details