Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000227.4 | Intron | NP_000218.3 | |||
NM_001127717.2 | Intron | NP_001121189.2 | |||
NM_001127718.2 | Intron | NP_001121190.2 | |||
NM_001302996.1 | Intron | NP_001289925.1 | |||
NM_198129.2 | Intron | NP_937762.2 | |||
XM_011525978.2 | Intron | XP_011524280.1 | |||
XM_011525979.2 | Intron | XP_011524281.1 | |||
XM_011525980.2 | Intron | XP_011524282.1 | |||
XM_011525981.2 | Intron | XP_011524283.1 | |||
XM_011525982.2 | Intron | XP_011524284.1 | |||
XM_017025743.1 | Intron | XP_016881232.1 | |||
XM_017025744.1 | Intron | XP_016881233.1 |