Product Details

SNP ID
rs16908498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92713070 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGGCACCTGAGACCGTCTCTACGC[A/G]AGGAATTAAGGAAGCAAATATAATA
Phenotype
MIM: 609797
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BICD2 PubMed Links

Gene Details

Gene
BICD2
Gene Name
BICD cargo adaptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003800.1 3039 UTR 3 NP_001003800.1
NM_015250.3 3039 UTR 3 NP_056065.1
XM_017014551.1 3039 UTR 3 XP_016870040.1

View Full Product Details