Product Details

SNP ID
rs17826038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43726529 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACAAACAGCCTCATGTTCCTCCA[C/G]ATTTTTGATGGGGACCACGTGGCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GTSF1L PubMed Links

Gene Details

Gene
GTSF1L
Gene Name
gametocyte specific factor 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008901.1 474 Missense Mutation CTG,GTG L56V NP_001008901.1
NM_176791.3 474 Missense Mutation CTG,GTG L56V NP_789761.1
XM_005260298.4 474 Missense Mutation CTG,GTG L56V XP_005260355.1
Gene
LOC101927200
Gene Name
uncharacterized LOC101927200
There are no transcripts associated with this gene.

View Full Product Details