Product Details

SNP ID
rs16920295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:56071605 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGAACTGTGAGACTTCAGTAGAC[A/C]ATGGATTCAGACTAGGAACACAGGA
Phenotype
MIM: 603682
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC105375847 PubMed Links

Gene Details

Gene
LOC105375847
Gene Name
uncharacterized LOC105375847
There are no transcripts associated with this gene.

Gene
RPS20
Gene Name
ribosomal protein S20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001023.3 Intron NP_001014.1
NM_001146227.1 Intron NP_001139699.1
Gene
SNORD54
Gene Name
small nucleolar RNA, C/D box 54
There are no transcripts associated with this gene.

View Full Product Details