Product Details

SNP ID
rs17857117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50185584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAAGGGGGCCACATCGATGATG[C/G]GCAGGCGGGAGGTCTTGGTGGTTTT
Phenotype
MIM: 120150 MIM: 600119
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COL1A1 PubMed Links

Gene Details

Gene
COL1A1
Gene Name
collagen type I alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000088.3 4250 Missense Mutation CCC,CGC P1438R NP_000079.2
XM_005257058.3 4250 Missense Mutation CCC,CGC P1348R XP_005257115.2
XM_005257059.4 4250 Missense Mutation CCC,CGC P1132R XP_005257116.2
XM_011524341.1 4250 Missense Mutation CCC,CGC P1372R XP_011522643.1
Gene
SGCA
Gene Name
sarcoglycan alpha
There are no transcripts associated with this gene.

View Full Product Details