Product Details

SNP ID
rs17078109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116601344 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGTCAAAACATATCTAAATGTGG[C/T]AGAATCATGAAATTAAATTGAATTA
Phenotype
MIM: 612647
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RSPH4A PubMed Links

Gene Details

Gene
RSPH4A
Gene Name
radial spoke head 4 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010892.2 3161 Intron NP_001010892.1
NM_001161664.1 3161 Intron NP_001155136.1
XM_017010826.1 3161 UTR 5 XP_016866315.1
Gene
RWDD1
Gene Name
RWD domain containing 1
There are no transcripts associated with this gene.

View Full Product Details