Product Details

SNP ID
rs11542013
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.1:151983282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTGCCATCTCTACACTGGTCCA[A/G]GTCCTTCATTATTTTGTCCACAGCC
Phenotype
MIM: 114085
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
S100A10 PubMed Links
Additional Information
For this assay, SNP(s) [rs146684690] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
S100A10
Gene Name
S100 calcium binding protein A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002966.2 683 Silent Mutation NP_002957.1

View Full Product Details