Product Details

SNP ID
rs11544936
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:45511358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGCTTGGAGAAATATTCTTTGC[C/T]CTTTTGGACATCAGGCTTGATGGTA
Phenotype
MIM: 609831 MIM: 176763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMACHC PubMed Links

Gene Details

Gene
MMACHC
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015506.2 912 Intron NP_056321.2
XM_005270724.4 912 Intron XP_005270781.1
XM_011541204.2 912 Intron XP_011539506.1
Gene
PRDX1
Gene Name
peroxiredoxin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202431.1 912 Missense Mutation AGC,GGC S191G NP_001189360.1
NM_002574.3 912 Missense Mutation AGC,GGC S191G NP_002565.1
NM_181696.2 912 Missense Mutation AGC,GGC S191G NP_859047.1
NM_181697.2 912 Intron NP_859048.1

View Full Product Details