Product Details

SNP ID
rs11549839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:4781669 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCGCCGCCATCCTGCGAGAGATGT[C/G]CCCCTCTGACAGCCTCAGCCTGGCC
Phenotype
MIM: 613653
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AP5Z1 PubMed Links

Gene Details

Gene
AP5Z1
Gene Name
adaptor related protein complex 5 zeta 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014855.2 366 Missense Mutation TCC,TGC S94C NP_055670.1
XM_017012864.1 366 Intron XP_016868353.1
Gene
MIR4656
Gene Name
microRNA 4656
There are no transcripts associated with this gene.

View Full Product Details