Product Details

SNP ID
rs9643863
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39462236 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATCAGTTCTATAATGTTAAAAT[A/G]CCTGTACCACACAAAGCAATTTACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM3A PubMed Links

Gene Details

Gene
ADAM3A
Gene Name
ADAM metallopeptidase domain 3A (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details