Product Details

SNP ID
rs11537875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:140511541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTAAAGACAAGGGCGGGGAGCA[A/G]CGGCTCATCTCGCTGCCTCTATCCC
Phenotype
MIM: 606229 MIM: 607268
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGO2 PubMed Links

Gene Details

Gene
AGO2
Gene Name
argonaute 2, RISC catalytic component
There are no transcripts associated with this gene.

Gene
CHRAC1
Gene Name
chromatin accessibility complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017444.5 244 Silent Mutation CAA,CAG Q14Q NP_059140.1

View Full Product Details