Product Details

SNP ID
rs11557143
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:32587078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTCAGAAGAGAACTCGGAAGGT[A/G]GACTTCATGTTGATTTAGCTCAAAT
Phenotype
MIM: 609641
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
EIF3M PubMed Links

Gene Details

Gene
EIF3M
Gene Name
eukaryotic translation initiation factor 3 subunit M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307929.1 254 Intron NP_001294858.1
NM_006360.5 254 Missense Mutation AGA,GGA R37G NP_006351.2
XM_011519840.2 254 UTR 5 XP_011518142.1

View Full Product Details