Product Details

SNP ID
rs11554936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48616784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGTTATGGTCCCCACAACCACG[A/G]CCGTCTTGTTTTCCCGGCCAGGAAG
Phenotype
MIM: 604179 MIM: 609932 MIM: 607092
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM83E PubMed Links

Gene Details

Gene
FAM83E
Gene Name
family with sequence similarity 83 member E
There are no transcripts associated with this gene.

Gene
RPL18
Gene Name
ribosomal protein L18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000979.3 514 Missense Mutation GCC,GTC A80V NP_000970.1
NM_001270490.1 514 Missense Mutation GCC,GTC A51V NP_001257419.1
Gene
SPACA4
Gene Name
sperm acrosome associated 4
There are no transcripts associated with this gene.

Gene
SPHK2
Gene Name
sphingosine kinase 2
There are no transcripts associated with this gene.

View Full Product Details