Product Details
- SNP ID
-
hCV43904722
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:62246697 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGGTGCTGGGTGAAGAGAGCTGC[G/T]CCCCTGTCCCTCAGCCCCTGCCCCT
- Phenotype
-
MIM: 606731
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OSBPL2
PubMed Links
Gene Details
- Gene
- OSBPL2
- Gene Name
- oxysterol binding protein like 2
There are no transcripts associated with this gene.
View Full Product Details