Product Details

SNP ID
rs13441234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23701436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTGTGACTATTTCATGCCAGAG[G/T]TGCTTGTTGACACTGTGATTTCAAT
Phenotype
MIM: 300862
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACOT9 PubMed Links

Gene Details

Gene
ACOT9
Gene Name
acyl-CoA thioesterase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033583.2 3823 Intron NP_001028755.2
NM_001037171.1 3823 Intron NP_001032248.1
XM_005274472.2 3823 UTR 3 XP_005274529.1

View Full Product Details