Product Details
- SNP ID
-
rs5743734
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:12871690 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTCTGCTCTGTGATCTTGAAAGT[C/T]GCTTGGAGGAACAGCTGAGTGCATG
- Phenotype
-
MIM: 300365
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TLR7
PubMed Links
Gene Details
- Gene
- TLR7
- Gene Name
- toll like receptor 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016562.3 |
|
Intron |
|
|
NP_057646.1 |
View Full Product Details