Product Details

SNP ID
rs6926874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149510138 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAGTATGGCCTGAAAACGGGAT[A/G]AGGCATGAATGATGGCAATCTTTAA
Phenotype
MIM: 607609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPIL4 PubMed Links

Gene Details

Gene
PPIL4
Gene Name
peptidylprolyl isomerase like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139126.3 Intron NP_624311.1

View Full Product Details