Product Details

SNP ID
rs11556304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58257831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACTTCAGATCTAACTTTGGCTA[C/T]AACATTCCACTAAAAGTAAGTTGAT
Phenotype
MIM: 176843
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMA3 PubMed Links

Gene Details

Gene
PSMA3
Gene Name
proteasome subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002788.3 431 Silent Mutation TAC,TAT Y105Y NP_002779.1
NM_152132.2 431 Silent Mutation TAC,TAT Y105Y NP_687033.1
Gene
PSMA3-AS1
Gene Name
PSMA3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details