Product Details
- SNP ID
-
rs9324211
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:97202133 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAGGACAGAGGTTGATTCTAACA[C/G]TGCTCGAATGTATGGAAATCTTTGT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD36
PubMed Links
Gene Details
- Gene
- ANKRD36
- Gene Name
- ankyrin repeat domain 36
View Full Product Details