Product Details

SNP ID
rs28648904
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:151724314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCTTCTGACCTCTTTTCCTGGG[G/T]GGAGCTTGCAGAGGGTTTAGTGTTT
Phenotype
MIM: 612678
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CELF3 PubMed Links

Gene Details

Gene
CELF3
Gene Name
CUGBP, Elav-like family member 3
There are no transcripts associated with this gene.

Gene
RIIAD1
Gene Name
regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144956.1 Intron NP_001138428.1

View Full Product Details