Product Details

SNP ID
rs28371575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122995288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAAAATTCTCCACCTCAGCCAG[G/A]GAAAGATGTGATGGGTGAAAGAAGA
Phenotype
MIM: 604867
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
TAS2R16 PubMed Links

Gene Details

Gene
TAS2R16
Gene Name
taste 2 receptor member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016945.2 413 Missense Mutation CCC,CTC P116L NP_058641.1

View Full Product Details