Product Details

SNP ID
rs2381004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33395121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAACTCGGCCAGGAACTCTCGC[A/G]CCATCTTCCTCTGCAGTATTTCCTG
Phenotype
MIM: 602974
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AQP7 PubMed Links

Gene Details

Gene
AQP7
Gene Name
aquaporin 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170.2 Intron NP_001161.1
NM_001318156.1 Intron NP_001305085.1
NM_001318157.1 Intron NP_001305086.1
NM_001318158.1 Intron NP_001305087.1
XM_005251453.3 Intron XP_005251510.1
XM_006716765.3 Intron XP_006716828.1
XM_011517866.2 Intron XP_011516168.1
XM_011517867.2 Intron XP_011516169.1
XM_017014700.1 Intron XP_016870189.1
XM_017014701.1 Intron XP_016870190.1
XM_017014702.1 Intron XP_016870191.1
XM_017014703.1 Intron XP_016870192.1
XM_017014704.1 Intron XP_016870193.1
XM_017014705.1 Intron XP_016870194.1
XM_017014706.1 Intron XP_016870195.1

View Full Product Details