Product Details

SNP ID
rs28702860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110722630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGATCACAAAGAAGACCTTGAT[C/T]TGGTACATCAGTTGGTCGCATAATT
Phenotype
MIM: 176914
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1CC PubMed Links

Gene Details

Gene
PPP1CC
Gene Name
protein phosphatase 1 catalytic subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244974.1 781 Missense Mutation AAT,GAT N197D NP_001231903.1
NM_002710.3 781 Missense Mutation AAT,GAT N197D NP_002701.1
XM_011538504.2 781 Missense Mutation AAT,GAT N197D XP_011536806.1
XM_011538505.2 781 Missense Mutation AAT,GAT N197D XP_011536807.1

View Full Product Details