Product Details

SNP ID
rs28587788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75730458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGCAAAATGACAGGGTCATTTA[C/G]CTTGGGATTGACCTTTGGATGAAGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNM1P35 PubMed Links

Gene Details

Gene
DNM1P35
Gene Name
dynamin 1 pseudogene 35
There are no transcripts associated with this gene.

Gene
ODF3L1
Gene Name
outer dense fiber of sperm tails 3 like 1
There are no transcripts associated with this gene.

View Full Product Details