Product Details

SNP ID
rs28670224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60006397 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGCTCCCAGTAGACGCTCTGGG[C/T]TTCCTACCCTGCCCAGACACTCTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101927755 PubMed Links

Gene Details

Gene
LOC101927755
Gene Name
uncharacterized LOC101927755
There are no transcripts associated with this gene.

Gene
TBC1D3P1-DHX40P1
Gene Name
TBC1D3P1-DHX40P1 readthrough, transcribed pseudogene
There are no transcripts associated with this gene.

View Full Product Details