Product Details

SNP ID
rs3209255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36152403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGGGGCCTCACCCGAAGGGCC[T/A]GCATTCTGCCTTGTCCTCTTCCGCC
Phenotype
MIM: 114170 MIM: 123995
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
CAPNS1 PubMed Links

Gene Details

Gene
CAPNS1
Gene Name
calpain small subunit 1
There are no transcripts associated with this gene.

Gene
COX7A1
Gene Name
cytochrome c oxidase subunit 7A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001864.3 467 Missense Mutation CAG,CTG Q2L NP_001855.1

View Full Product Details